missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid, TRC

Product Code. 30419371
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419371 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419371 Supplier Toronto Research Chemicals Supplier No. TRCS699445500MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Sulfosalicylic Acid
CAS 97-05-2
Molecular Formula C7 H6 O6 S
Quantity 500 mg
Synonym Benzoic acid','2-hydroxy-5-sulfo-, Salicylic acid','5-sulfo- (8CI), 2-Hydroxy-5-sulfobenzoic acid, 3-Carboxy-4-hydroxybenzenesulfonic acid, 5-(Hydroxysulfonyl)salicylic acid, 5-Sulfosalicyclic acid, 5-Sulfosalicylic acid, 5-Sulphosalicylic acid, FD, NSC 190
InChI Formula InChI=1S/C7H6O6S/c8-6-2-1-4(14(11,12)13)3-5(6)7(9)10/h1-3,8H,(H,9,10)(H,11,12,13)
SMILES OC(=O)c1cc(ccc1O)S(=O)(=O)O
IUPAC Name 2-hydroxy-5-sulfobenzoic acid
Molecular Weight (g/mol) 218.184
Formula Weight 217.989
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.