missing translation for 'onlineSavingsMsg'
Learn More

Sulfosalicylic Acid Dihydrate (Fine White Crystals), Fisher BioReagents

Product Code. 11472944
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11472944 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11472944 Supplier Fisher BioReagents Supplier No. BP177500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sulfosalicylic Acid is used for fixing proteins in agarose and polyacrylamide gels.
TRUSTED_SUSTAINABILITY

Specifications

CAS 5965-83-3
Melting Point 106°C
Color White
pH 0.5
Ignition Residue 0.1% max.
Assay Percent Range 99.0 to 101.0 %
Packaging Glass Bottle
Molecular Formula C9H14O6S
Linear Formula HOC6H3(COOH)SO3H·2H2O
MDL Number MFCD00007508,MFCD00149540
Quantity 500 g
Merck Index 15, 9094
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
Solubility Information 0.02% max. Insoluble Matter
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
IUPAC Name 2-hydroxy-5-sulfobenzoic acid;dihydrate
Molecular Weight (g/mol) 250.27
PubChem CID 2723734
Formula Weight 254.22
Percent Purity 99.0 to 101.0%
Physical Form Powder or Solid
Chemical Name or Material Sulfosalicylic Acid Dihydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Sulfosalicylic acid dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, 5-Sulfosalicylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.