missing translation for 'onlineSavingsMsg'
Learn More

Sulforhodamine B sodium salt, MedChemExpress

Product Code. 16498916
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498916 100 mg 100mg
16482429 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498916 Supplier Medchem Express Supplier No. HYD0974

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sulforhodamine B sodium salt is a fluorescent dye with uses spanning from laser-induced fluorescence (LIF) to the quantification of cellular proteins of cultured cells.

Sulforhodamine B sodium salt is a fluorescent dye with uses spanning from laser-induced fluorescence (LIF) to the quantification of cellular proteins of cultured cells.

Specifica

Chemical Name or Material Sulforhodamine B sodium salt
CAS 3520-42-1
Purity Grade Notes Research
Molecular Formula C27H29N2NaO7S2
Synonym Acid Red 52 Kiton Red 620
Solubility Information DMSO : 50 mg/mL (86.11 mM; Need ultrasonic) ∣H2O : 20 mg/mL (34.44 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(C2=C3C=CC(N(CC)CC)=CC3=[O+]C4=C2C=CC(N(CC)CC)=C4)C(S(=O)([O-])=O)=C1)(O[Na])=O
Molecular Weight (g/mol) 580.65
Formula Weight 580.65
Percent Purity 98.88%
Grade Research
Color Brown
Quantity 100 mg
Physical Form Powder
Vedi altri risultati Mostra meno risultati

Research purposes only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.