missing translation for 'onlineSavingsMsg'
Learn More

Sulforhodamine B, MP Biomedicals™

Product Code. 11492752 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11492752 25 g 25g
11472752 1 g 1g
11482752 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 11492752

Brand: MP Biomedicals 0215247925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 3520-42-1
Molecular Formula C29H33N2NaO7S2
Molecular Weight (g/mol) 608.70
MDL Number MFCD00010180
InChI Key XJENLUNLXRJLEZ-UHFFFAOYSA-M
Synonym Acid Red 52, Phloxinrhodamine
PubChem CID 131852807
IUPAC Name sodium 3,6-bis(diethylamino)-9-(2,4-disulfonatophenyl)-2,7-dimethyl-10λ⁴-xanthen-10-ylium
SMILES [Na+].CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C2C=C1C

Specifications

Color Brown-Purple
Quantity 25 g
Formula Weight 580.66
Physical Form Powder
Chemical Name or Material Sulforhodamine B
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.