missing translation for 'onlineSavingsMsg'
Learn More

Sulforhodamine 101, TRC

Product Code. 30420486
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30420486

Brand: Toronto Research Chemicals TRCS68912025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulforhodamine 101
CAS 60311-02-6
Molecular Formula C31H30N2O7S2
Quantity 25 mg
Synonym Sulforhodamine 640, LC 6600, SR 101, 9-(2,4-Disulfophenyl)-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-Xantheno[2,3,4-ij:5,6,7-i'j']diquinolizin-18-ium Hydroxide Inner Salt
InChI Formula InChI=1S/C31H30N2O7S2/c34-41(35,36)20-9-10-21(26(17-20)42(37,38)39)27-24-15-18-5-1-11-32-13-3-7-22(28(18)32)30(24)40-31-23-8-4-14-33-12-2-6-19(29(23)33)16-25(27)31/h9-10,15-17H,1-8,11-14H2,(H-,34,35,36,37,38,39)
SMILES OS(=O)(=O)c1ccc(c2c3cc4CCCN5CCCc(c45)c3[o+]c6c7CCCN8CCCc(cc26)c78)c(c1)S(=O)(=O)[O-]
Molecular Weight (g/mol) 606.71
Formula Weight 606.1494
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.