missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulforhodamine 101, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | Sulforhodamine 101 |
| CAS | 60311-02-6 |
| Molecular Formula | C31H30N2O7S2 |
| Quantity | 500 mg |
| Synonym | Sulforhodamine 640, LC 6600, SR 101, 9-(2,4-Disulfophenyl)-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-Xantheno[2,3,4-ij:5,6,7-i'j']diquinolizin-18-ium Hydroxide Inner Salt |
| InChI Formula | InChI=1S/C31H30N2O7S2/c34-41(35,36)20-9-10-21(26(17-20)42(37,38)39)27-24-15-18-5-1-11-32-13-3-7-22(28(18)32)30(24)40-31-23-8-4-14-33-12-2-6-19(29(23)33)16-25(27)31/h9-10,15-17H,1-8,11-14H2,(H-,34,35,36,37,38,39) |
| SMILES | OS(=O)(=O)c1ccc(c2c3cc4CCCN5CCCc(c45)c3[o+]c6c7CCCN8CCCc(cc26)c78)c(c1)S(=O)(=O)[O-] |
| Molecular Weight (g/mol) | 606.71 |
| Formula Weight | 606.1494 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction