missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sulforhodamine 101 (free acid), 99%, pure, laser grade

Product Code. 10075950 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Unit Size:
100mg
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10075950 500 mg 500mg
10686224 100 mg 100mg
11926941 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10075950 Supplier Thermo Scientific Chemicals Supplier No. 419945000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60311-02-6 | C31H29ClN2O6S2 | 625.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60311-02-6
Molecular Formula C31H29ClN2O6S2
Molecular Weight (g/mol) 625.15
MDL Number MFCD00012407
InChI Key MPLHNVLQVRSVEE-UHFFFAOYSA-N
Synonym sulforhodamine 101, sulforhodamine 101; sulforhodamine 640, unii-fx0es3271v, texas red, sulforhodamine 101 acid chloride, 2-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-pyrido 3,2,1-ij quinolizino 1',9':6,7,8 chromeno 2,3-f quinolin-4-ium-9-yl-5-sulfobenzenesulfonate, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9$l^ 5 ,23-diazaheptacyclo 17.7.1.1^ 5,9 .0^ 2,17 .0^ 4,15 .0^ 23,27 .0^ 13,28 octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9??,23-diazaheptacyclo 17.7.1.1?,?.0 2 , 1 ?.0?, 1 ?.0 2 3 , 2 ?.0 1 3 , 2 ? octacosa-1 27 ,2 17 ,4,9 28 ,13,15,18-heptaen-9-ylium, 16-4-sulfo-2-sulfonatophenyl-3-oxa-9lambda,23-diazaheptacyclo 17.7.1.1,.0 2 , 1 .0, 1 .0 2 3 , 2 .0 1 3 , 2 octacosa-1,4,9 28 ,13,15,17,19 27-heptaen-9-ylium, 9-2,4-disulfophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium inner salt, hydrogen 9-2,4-disulphonatophenyl-2,3,6,7,12,13,16,17-octahydro-1h,5h,11h,15h-xantheno 2,3,4-ij:5,6,7-i'j' diquinolizin-18-ium
PubChem CID 122180
ChEBI CHEBI:52274
IUPAC Name 16-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3λ⁴-oxa-9,23-diazaheptacyclo[17.7.1.1⁵,⁹.0²,¹⁷.0⁴,¹⁵.0²³,²⁷.0¹³,²⁸]octacosa-1,3,5(28),13,15,17,19(27)-heptaen-3-ylium
SMILES [O-]S(=O)(=O)C1=CC(=CC=C1C1=C2C=C3CCCN4CCCC(C2=[O+]C2=C5CCCN6CCCC(C=C12)=C56)=C34)S(Cl)(=O)=O

Specifications

Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Formula Weight 606.71
Percent Purity 99%
Grade Pure
Chemical Name or Material Sulforhodamine 101 (free acid), (free acid)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.