missing translation for 'onlineSavingsMsg'
Learn More

Sulforaphane, MedChemExpress

Product Code. 16471987
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16471987

Brand: Medchem Express HY13755

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sulforaphane is an isothiocyanate present naturally in widely consumed vegetables. Sulforaphane increases tumor suppressor protein transcription and inhibits histone deacetylase activity. Sulforaphane activates Nrf2 and inhibits high glucose-induced progression of pancreatic cancer via AMPK dependent signaling. Sulforaphane has shown anti-cancer and anti-inflammatory activities.

Sulforaphane is an isothiocyanate present naturally in widely consumed vegetables. Sulforaphane increases tumor suppressor protein transcription and inhibits histone deacetylase activity. Sulforaphane activates Nrf2 and inhibits high glucose-induced progression of pancreatic cancer via AMPK dependent signaling. Sulforaphane has shown anti-cancer and anti-inflammatory activities.

Specifications

Chemical Name or Material Sulforaphane
Physical Form Liquid
CAS 4478-93-7
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H11NOS2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 62.5 mg/mL (352.53 mM)
SMILES O=S(CCCCN=C=S)C
Molecular Weight (g/mol) 177.29
Formula Weight 177.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.