missing translation for 'onlineSavingsMsg'
Learn More

Sulforaphane, MedChemExpress

Product Code. 16485838
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485838 10 mg 10mg
16471987 50 mg 50mg
16433386 25 mg 25mg
16406316 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485838 Supplier Medchem Express Supplier No. HY13755
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sulforaphane is an isothiocyanate present naturally in widely consumed vegetables. Sulforaphane increases tumor suppressor protein transcription and inhibits histone deacetylase activity. Sulforaphane activates Nrf2 and inhibits high glucose-induced progression of pancreatic cancer via AMPK dependent signaling. Sulforaphane has shown anti-cancer and anti-inflammatory activities.

Sulforaphane is an isothiocyanate present naturally in widely consumed vegetables. Sulforaphane increases tumor suppressor protein transcription and inhibits histone deacetylase activity. Sulforaphane activates Nrf2 and inhibits high glucose-induced progression of pancreatic cancer via AMPK dependent signaling. Sulforaphane has shown anti-cancer and anti-inflammatory activities.

Specifications

Chemical Name or Material Sulforaphane
Physical Form Liquid
CAS 4478-93-7
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C6H11NOS2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 62.5 mg/mL (352.53 mM)
SMILES O=S(CCCCN=C=S)C
Molecular Weight (g/mol) 177.29
Formula Weight 177.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.