missing translation for 'onlineSavingsMsg'
Learn More

Sulfonazo III Sodium Salt, Honeywell™ Fluka™

Product Code. 15628470 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 15628470

Brand: Honeywell-Fluka 861705G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Titration Indicator (for SO4)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 164581-28-6
Molecular Formula C22H12N4Na4O14S4
MDL Number MFCD00003942
UN Number NONH for all modes of transport
Beilstein 1900148
InChI Key OWVXGCUJHKOBMQ-SGLNGUFESA-J
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4S(=O)(=O)[O-])C(=C3)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
IUPAC Name tetrasodium;(3Z,6E)-4,5-dioxo-3,6-bis[(2-sulfonatophenyl)hydrazinylidene]naphthalene-2,7-disulfonate
Molecular Weight (g/mol) 776.551
PubChem CID 71299731
Formula Weight 776.57g/mol
Packaging Poly bottle
Quantity 5 g
Physical Form Powder
Chemical Name or Material Sulfonazo III sodium salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.