missing translation for 'onlineSavingsMsg'
Learn More

Sulfonazo III salt

Product Code. 11478723
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478723 5 g 5g
11488723 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11478723 Supplier Thermo Scientific Alfa Aesar Supplier No. A10486.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68504-35-8 | C22H16N4Na4O14S4 | 780.583 g/mol

Sulfonazo III salt is used as spectrophotometric reagent for alkaline earth metals and indicator for sulfate titrations. In general it is used as an indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfonazo III salt is used as spectrophotometric reagent for alkaline earth metals and indicator for sulfate titrations. In general it is used as an indicator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in a cool, dry, well-ventilated area.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68504-35-8
Molecular Formula C22H16N4Na4O14S4
Molecular Weight (g/mol) 780.583
MDL Number MFCD00003942
InChI Key WHVMUCGHHVLOCE-FMNKGCAFSA-N
Synonym Tetrasodium 4,5-dihydroxy-3,6-bis[(2-sulfonatophenyl)azo]naphthalene-2,7-disulfonate
PubChem CID 131857569
IUPAC Name (3E,6E)-4,5-dioxo-3,6-bis[(2-sulfophenyl)hydrazinylidene]naphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC=C(C(=C1)NN=C2C(=CC3=C(C2=O)C(=O)C(=NNC4=CC=CC=C4S(=O)(=O)O)C(=C3)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na].[Na].[Na]

Specifications

Melting Point ≥300°C
Quantity 5 g
Beilstein 1900148
Solubility Information Soluble in water.
Formula Weight 776.57
Chemical Name or Material Sulfonazo III salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.