missing translation for 'onlineSavingsMsg'
Learn More

Sulfolane, 99%

Product Code. 11415543
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
This item is not returnable. View return policy

Product Code. 11415543

Brand: Thermo Scientific Alfa Aesar A13466.0I

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Miscible with water, acetone, toluene | CAS: 126-33-0 | C4H8O2S | 120.17 g/mol

Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfolane is used as an industrial solvent utilized for the extraction of aromatic hydrocarbons from hydrocarbon mixtures and to purify natural gas. Further, it is used in refineries and the petrochemical industry.

Solubility
Miscible with alcohol, water, acetone, toluene and dilute mineral acids. Slightly miscible with octanes, olefins and naphthenes.

Notes
Miscible with water, acetone, tolueneIncompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-33-0
Molecular Formula C4H8O2S
Molecular Weight (g/mol) 120.17
MDL Number MFCD00005484
InChI Key HXJUTPCZVOIRIF-UHFFFAOYSA-N
Synonym sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone
PubChem CID 31347
ChEBI CHEBI:74794
SMILES O=S1(=O)CCCC1

Specifications

Melting Point 25°C to 28°C
Density 1.261
Boiling Point 285°C
Flash Point 166°C (330°F)
Assay Percent Range 99%
Odor Weak, Characteristic
Refractive Index 1.4825
Quantity 5000 g
Beilstein 107765
Sensitivity Hygroscopic
Merck Index 14,8955
Solubility Information Miscible with alcohol,water,acetone,toluene and dilute mineral acids. Slightly miscible with octanes,olefins and naphthenes.
IUPAC Name thiolane 1,1-dioxide
Formula Weight 120.17
Percent Purity 99%
Chemical Name or Material Sulfolane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Sulfolane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H302-Harmful if swallowed.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetramethylene sulfone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.