missing translation for 'onlineSavingsMsg'
Learn More

Sulfobromophthalein sodium hydrate

Product Code. 10195103 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10195103 100 g Glass Bottle 100g
10419310 5 g Glass Bottle 5g
10712411 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10195103 Supplier Thermo Scientific Acros Supplier No. 173401000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 123359-42-2 | C20H8Br4Na2O10S2 | 837.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123359-42-2
Molecular Formula C20H8Br4Na2O10S2
Molecular Weight (g/mol) 837.99
MDL Number MFCD16556312
InChI Key GHAFORRTMVIXHS-UHFFFAOYSA-L
Synonym disodium 2-hydroxy-5-1z-4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene 2,3,4,5-tetrabromo-6-carboxyphenyl methyl benzenesulfonate tetrahydrate
PubChem CID 16211419
IUPAC Name disodium;2-hydroxy-5-[(E)-(4-oxo-3-sulfonatocyclohexa-2,5-dien-1-ylidene)-(2,3,4,5-tetrabromo-6-carboxyphenyl)methyl]benzenesulfonate
SMILES [Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O

Specifications

Color White
Infrared Spectrum Authentic
Packaging Glass Bottle
Quantity 100 g
Solubility Information Solubility in water: soluble
Formula Weight 837.76
Percent Purity ≥95%
Physical Form Crystalline Powder
Chemical Name or Material Sulfobromophthalein sodium hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.