missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfo-SMCC sodium, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Sulfo-SMCC sodium is a commonly used hetero-bifunctional, noncleavable ADC crosslinker bearing N-hydroxysuccinimide (NHS) ester and maleimide groups to react with primary amines and sulfhydryl groups, respectively.
Specifications
Specifications
| Chemical Name or Material | Sulfo-SMCC sodium |
| CAS | 92921-24-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H17N2NaO9S |
| Quantity | 50 mg |
| Solubility Information | DMSO : 125 mg/mL (286.45 mM; Need ultrasonic) ∣H2O : 5 mg/mL (11.46 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(C1CCC(CN2C(C=CC2=O)=O)CC1)ON3C(C(S(=O)(O[Na])=O)CC3=O)=O |
| Molecular Weight (g/mol) | 436.37 |
| Formula Weight | 436.37 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction