missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Sulfo-NHS-LC-Biotin

Product Code. 18715606
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18715606 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18715606 Supplier Tocris Bioscience™ Supplier No. 7302/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Amine reactive biotinylation reagent, cell impermeable

Sulfo-NHS-LC-Biotin is a water-soluble amine reactive biotinylation reagent used to attach biotin to primary amines. Commonly used to label proteins and antibodies. Does not penetrate membrane. Reactive in pH 7.0-9.0, solubility 10 mg/mL in water and DMF.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium 2,5-dioxo-1-((6-(5-((3 AS,4 S,6 aR)-2-oxohexahydro-1 H-thieno[3,4-d]imidazol-4-yl)pentanamido)hexanoyl)oxy)pyrrolidine-3-sulfonate
CAS 191671-46-2
Quantity 50 mg
Target General Laboratory Research Reagent
Molecular Formula C20H29N4NaO9S2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.