missing translation for 'onlineSavingsMsg'
Learn More

WTW™ Sulfite Cell Test Kit

Product Code. 11791093
Click to view available options
No. of Tests:
25
Unit Size:
Pack of 25
This item is not returnable. View return policy

Product Code. 11791093

Brand: WTW™ 250416

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

With certificate for optimum precision.

  • Convenient and cost-effective
  • Assured quality by AQA/IQC
  • For infrequent or very large sample throughput in the laboratory
  • Sample adding, inserting, measuring and reading
  • Reaction cuvette with barcode or method selection

TRUSTED_SUSTAINABILITY

Specifications

Analytes Ellman’s reagent
For Use With (Application) For waste water, boiler water, foodstuffs
For Use With (Equipment) PhotoLab™ S12, PhotoLab™ spectral
Includes Certificates
No. of Tests 25
Range 1.0 - 20.0mg/I SO₃
Sufficient For 25 Tests
Test Method Photometric
Type Sulfite (SO3) cell test kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.