missing translation for 'onlineSavingsMsg'
Learn More

Sulfisoxazole, MedChemExpress

Product Code. 16428236
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16428236 100 mg 100mg
16402106 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16428236 Supplier Medchem Express Supplier No. HYB0323
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Sulfisoxazole (Sulfafurazole) is an endothelin receptor antagonist with IC50 values of 0.60 μM and 22 μM against endothelin receptor A and endothelin receptor B, respectively. Sulfisoxazole is a sulfonamide antibacterial with an oxazole substituent. Sulfisoxazole inhibits breast cancer exosome release by targeting endothelin receptor A.

Sulfisoxazole (Sulfafurazole) is an endothelin receptor antagonist with IC50 values of 0.60 μM and 22 μM against endothelin receptor A and endothelin receptor B, respectively. Sulfisoxazole is a sulfonamide antibacterial with an oxazole substituent. Sulfisoxazole inhibits breast cancer exosome release by targeting endothelin receptor A.

Specifications

Chemical Name or Material Sulfisoxazole
Physical Form Solid
CAS 127-69-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C11H13N3O3S
For Use With (Application) COVID-19-immunoregulation
Synonym Sulfafurazole
Solubility Information DMSO : ≥ 150 mg/mL (561.17 mM)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=C(C)C(C)=NO2)=O
Molecular Weight (g/mol) 267.3
Formula Weight 267.3
Percent Purity 99.95%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.