missing translation for 'onlineSavingsMsg'
Learn More

Sulfatinib, MedChemExpress

Product Code. 16495806
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495806 100 mg 100mg
16491229 1 mg 1mg
16447797 5 mg 5mg
16445078 10 mg 10mg
16411317 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495806 Supplier Medchem Express Supplier No. HY12297

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sulfatinib (HMPL-012) is a potent and highly selective tyrosine kinase inhibitor against VEGFR1/2/3, FGFR1 and CSF1R with IC50s of in a range of 1 to 24 nM.

Sulfatinib (HMPL-012) is a potent and highly selective tyrosine kinase inhibitor against VEGFR1/2/3, FGFR1 and CSF1R with IC50s of in a range of 1 to 24 nM.

Specifications

Chemical Name or Material Sulfatinib
CAS 1308672-74-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H28N6O3S
Quantity 100 mg
Synonym HMPL-012
Solubility Information DMSO : ≥ 100 mg/mL (208.08 mM)
SMILES O=S(CC1=CC=CC(NC2=NC=CC(OC3=CC4=C(NC(C)=C4)C=C3)=N2)=C1)(NCCN(C)C)=O
Molecular Weight (g/mol) 480.58
Formula Weight 480.58
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.