missing translation for 'onlineSavingsMsg'
Learn More

Sulfathiazole-d4, TRC

Product Code. 30426441
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426441 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30426441 Supplier Toronto Research Chemicals Supplier No. TRCS6990942.5MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfathiazole-d4
CAS 1020719-89-4
Purity Grade Notes HPLC
Molecular Formula C9 2H4 H5 N3 O2 S2
Quantity 2.5 mg
Synonym Sulfathiazole D4 (benzene D4)
InChI Formula InChI=1S/C9H9N3O2S2/c10-7-1-3-8(4-2-7)16(13,14)12-9-11-5-6-15-9/h1-6H,10H2,(H,11,12)/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2nccs2
IUPAC Name 4-amino-2,3,5,6-tetradeuterio-N-(1,3-thiazol-2-yl)benzenesulfonamide
Molecular Weight (g/mol) 259.34
Formula Weight 259.0387
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.