missing translation for 'onlineSavingsMsg'
Learn More

Sulfathiazole, 98+%

Product Code. 11469592
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469592 100 g 100g
11479592 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469592 Supplier Thermo Scientific Alfa Aesar Supplier No. A10727.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72-14-0 | C9H9N3O2S2 | 255.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

  • Mixture of hexanes where n-hexane content is ≥60%
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) and tested for GC/MS suitability
  • Also meets ACS specifications
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-14-0
Molecular Formula C9H9N3O2S2
Molecular Weight (g/mol) 255.31
MDL Number MFCD00005319
InChI Key JNMRHUJNCSQMMB-UHFFFAOYSA-N
Synonym sulfathiazole, sulphathiazole, sulfathiazol, sulfanilamidothiazole, 2-sulfanilamidothiazole, norsulfazole, thiazamide, 2-sulfonamidothiazole, norsulfasol, neostrepsan
PubChem CID 5340
ChEBI CHEBI:9337
IUPAC Name 4-amino-N-(1,3-thiazol-2-yl)benzenesulfonamide
SMILES NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1

Specifications

Melting Point 200°C to 203°C
Assay Percent Range 99%
Quantity 100 g
Merck Index 14,8943
Solubility Information Slightly soluble in water,deuterated dimethyl sulfoxide and ethanol. Soluble in acetone,dilute mineral acids,potassium hydroxide solution,sodium hydroxide solution,ammonia water,dimethyl sulphoxide and methanol.
Formula Weight 255.32
Percent Purity 99%
Chemical Name or Material Sulfathiazole
compliance-icons
Product Identifier
  • Sulfathiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-amino-N-2-thiazolylbenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.