missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Sulfatase-2/SULF2 Polyclonal antibody specifically detects Sulfatase-2/SULF2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Sulfatase-2/SULF2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.05 - 0.1 ug/ml |
| Formulation | PBS |
| Gene Alias | DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from the human Sulfatase-2/SULF2 protein. (Uniprot: Q8IWU5) |
| Purification Method | Antigen Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?