missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfatase-2/SULF2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£225.00 - £401.00
Specifications
| Antigen | Sulfatase-2/SULF2 |
|---|---|
| Dilution | Western Blot 0.05 - 0.1 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18370582
|
Novus Biologicals
NBP3-11910-25UG |
25 μg |
£225.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18313943
|
Novus Biologicals
NBP3-11910-50UG |
50 μg |
£401.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sulfatase-2/SULF2 Polyclonal antibody specifically detects Sulfatase-2/SULF2 in Human samples. It is validated for Western BlotSpecifications
| Sulfatase-2/SULF2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS | |
| 55959 | |
| IgG | |
| Antigen Affinity-purified |
| Western Blot 0.05 - 0.1 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2 | |
| Synthetic peptide derived from the human Sulfatase-2/SULF2 protein. (Uniprot: Q8IWU5) | |
| Primary | |
| Store at -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title