missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Sulfatase-2/SULF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30991-100ug
This item is not returnable.
View return policy
Description
Sulfatase-2/SULF2 Polyclonal antibody specifically detects Sulfatase-2/SULF2 in Human samples. It is validated for Western Blot
Specifications
| Sulfatase-2/SULF2 | |
| Polyclonal | |
| Western Blot : 0.05-0.1 μg/mL | |
| DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2 | |
| Synthetic peptide derived from the human Sulfatase-2/SULF2 protein. | |
| 100 μg | |
| Lipid and Metabolism | |
| 55959 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction