missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfatase-2/SULF2 Antibody (2B4), mFluor Violet 450 SE, Novus Biologicals™
Description
Sulfatase-2/SULF2 Monoclonal antibody specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | Sulfatase-2/SULF2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2B4 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2 |
| Host Species | Mouse |
| Immunogen | Partial recombinant human Sulfatase 2 isoform b [Accession# NP_940998]. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?