missing translation for 'onlineSavingsMsg'
Learn More

Sulfatase-2/SULF2 Antibody (2B4), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18406523 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406523 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406523 Supplier Novus Biologicals Supplier No. NBP136727JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sulfatase-2/SULF2 Monoclonal antibody specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sulfatase-2/SULF2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone 2B4
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2
Host Species Mouse
Immunogen Partial recombinant human Sulfatase 2 isoform b [Accession# NP_940998].
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 55959
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.