missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfatase-2/SULF2 Antibody (2B4), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-36727R
This item is not returnable.
View return policy
Description
Sulfatase-2/SULF2 Monoclonal specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Sulfatase-2/SULF2 | |
| Monoclonal | |
| DyLight 550 | |
| NP_940998 | |
| SULF2 | |
| Partial recombinant human Sulfatase 2 isoform b [Accession# NP_940998]. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 55959 | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 2B4 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Does not recognize human or mouse Sulfatase 1 or other family members. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction