missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfatase-2/SULF2 Antibody (2B4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-36727APC
This item is not returnable.
View return policy
Description
Sulfatase-2/SULF2 Monoclonal specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| Sulfatase-2/SULF2 | |
| Monoclonal | |
| APC | |
| DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF-2, sulfatase 2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Does not recognize human or mouse Sulfatase 1 or other family members. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| 2B4 | |
| Flow Cytometry | |
| SULF2 | |
| Partial recombinant human Sulfatase 2 isoform b [Accession# NP_940998]. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 55959 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction