missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine 3-Isomer, TRC

Product Code. 30412447
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412447 50 mg 50mg
30426329 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30412447 Supplier Toronto Research Chemicals Supplier No. TRCS69913050MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66364-71-4
Molecular Formula C18H14N4O5S
Molecular Weight (g/mol) 398.39
Synonym 2-Hydroxy-3-[2-[4-[(2-pyridinylamino)sulfonyl]phenyl]diazenyl]benzoic Acid, 2-Hydroxy-3-[[4-[(2-pyridinylamino)sulfonyl]phenyl]azo]benzoic Acid, 3-[[p-(2-Pyridylsulfamoyl)phenyl]azo]salicylic Acid, Sulfasalazine Impurity F
SMILES C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N/N=C/3\C=CC=C(C3=O)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C18H14N4O5S/c23-17-14(18(24)25)4-3-5-15(17)21-20-12-7-9-13(10-8-12)28(26,27)22-16-6-1-2-11-19-16/h1-11,23H,(H,19,22)(H,24,25)
Chemical Name or Material Sulfasalazine 3-Isomer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.