missing translation for 'onlineSavingsMsg'
Learn More

Sulfaquinoxaline-d4, TRC

Product Code. 30427549
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427549 10 mg 10mg
30426977 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30427549 Supplier Toronto Research Chemicals Supplier No. TRCS68903710MG
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1329652-02-9
Molecular Formula C14 2H4 H8 N4 O2 S
Molecular Weight (g/mol) 304.3603
Synonym Sulfaquinoxaline D4 (Sulfanilyl D4), 4-Amino-N-2-quinoxalinylbenzene-2,3,5,6-d4-sulfonamide, Benzene-2,3,5,6-d4-sulfonamide, 4-amino-N-2-quinoxalinyl-
IUPAC Name 4-amino-2,3,5,6-tetradeuterio-N-quinoxalin-2-ylbenzenesulfonamide
SMILES [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2cnc3ccccc3n2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H12N4O2S/c15-10-5-7-11(8-6-10)21(19,20)18-14-9-16-12-3-1-2-4-13(12)17-14/h1-9H,15H2,(H,17,18)/i5D,6D,7D,8D
Formula Weight 304.0932
Chemical Name or Material Sulfaquinoxaline sulfanilyl-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.