missing translation for 'onlineSavingsMsg'
Learn More

Sulfapyridine, 95%, Thermo Scientific Chemicals

Product Code. 15432152 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15432152

Brand: Thermo Scientific Acros 442851000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-83-2
Molecular Formula C11H11N3O2S
Molecular Weight (g/mol) 249.288
InChI Key GECHUMIMRBOMGK-UHFFFAOYSA-N
PubChem CID 5336
ChEBI CHEBI:132842
IUPAC Name 4-amino-N-pyridin-2-ylbenzenesulfonamide
SMILES C1=CC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Type Sulfapyridine
Melting Point 190.0°C
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Quantity 100 g
Formula Weight 249.29
Percent Purity 95%
Chemical Name or Material Sulfapyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.