missing translation for 'onlineSavingsMsg'
Learn More

Sulfanitran, TRC

Product Code. 30408972
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408972 1 g 1g
30417546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30408972 Supplier Toronto Research Chemicals Supplier No. TRCS6890401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfanitran
CAS 122-16-7
Molecular Formula C14 H13 N3 O5 S
Quantity 1 g
Synonym Acetamide','N-[4-[[(4-nitrophenyl)amino]sulfonyl]phenyl]- (9CI','ACI), Acetanilide','4'-[(p-nitrophenyl)sulfamoyl]- (6CI','7CI','8CI), N-[4-[[(4-Nitrophenyl)amino]sulfonyl]phenyl]acetamide (ACI), 4'-[(p-Nitrophenyl)sulfamoyl]acetanilide, N4-Acetyl-4'-nitrosulfanilanilide, N4-Acetyl-N1-(4-nitrophenyl)sulfanilamide, N4-Acetyl-N1-(p-nitrophenyl)sulfanilamide, NSC 217299, NSC 77120, Sulfanitran
InChI Formula InChI=1S/C14H13N3O5S/c1-10(18)15-11-4-8-14(9-5-11)23(21,22)16-12-2-6-13(7-3-12)17(19)20/h2-9,16H,1H3,(H,15,18)
SMILES CC(=O)Nc1ccc(cc1)S(=O)(=O)Nc2ccc(cc2)[N+](=O)[O-]
IUPAC Name N-[4-[(4-nitrophenyl)sulfamoyl]phenyl]acetamide
Molecular Weight (g/mol) 335.34
Formula Weight 335.0576
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.