missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfanitran, MedChemExpress
Description
Sulfanitran is an antibacterial and anticoccidial agent used in poultry feeds. Sulfanitran also is a multidrug resistance protein 2 (MRP2) stimulator that can increase the affinity of MRP2 for estradiol-17-β-D-glucuronide (E217βG).
Specifications
Specifications
| Chemical Name or Material | Sulfanitran |
| Physical Form | Powder |
| CAS | 122-16-7 |
| Color | Off-White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C14H13N3O5S |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Solubility Information | DMSO : ≥ 22 mg/mL (65.61 mM) |
| SMILES | CC(NC1=CC=C(S(=O)(NC2=CC=C([N+]([O-])=O)C=C2)=O)C=C1)=O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?