missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilic acid, 98+%

Product Code. p-7022866
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472498 250 g 250g
11462498 1000 g 1000g
11482498 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472498 Supplier Thermo Scientific Alfa Aesar Supplier No. A15825.30
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent for the detection of histidine. Intermediate for dyes and pharmaceuticals | CAS: 121-57-3 | C6H7NO3S | 173.186 g/mol

Reagent for the detection of histidine; intermediate for dyes and pharmaceuticalsSulfanilic acid is used as a reagent for chromatographic detection of histidine. It is also used as a chemical intermediate in organic synthesis. It is utilized in quantitative analysis of nitrate and nitrite ions. It plays an essential role in the manufacturing of azo dyes and in synthesis of sulfa drugs. It is used as a dopant for the chemical synthesis of polyaniline. It is used to prepare 2,4-diamino-6-p-sulphoanilinopyrimidine by reacting with 6-chloro-pyrimidine-2,4-diamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for the detection of histidine; intermediate for dyes and pharmaceuticalsSulfanilic acid is used as a reagent for chromatographic detection of histidine. It is also used as a chemical intermediate in organic synthesis. It is utilized in quantitative analysis of nitrate and nitrite ions. It plays an essential role in the manufacturing of azo dyes and in synthesis of sulfa drugs. It is used as a dopant for the chemical synthesis of polyaniline. It is used to prepare 2,4-diamino-6-p-sulphoanilinopyrimidine by reacting with 6-chloro-pyrimidine-2,4-diamine.

Solubility
Slightly soluble in water. Insoluble in ethanol, ether, caustic soda and sodium carbonate.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-57-3
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
MDL Number MFCD00007886
InChI Key HVBSAKJJOYLTQU-UHFFFAOYSA-N
Synonym sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
PubChem CID 8479
ChEBI CHEBI:27500
IUPAC Name 4-aminobenzenesulfonic acid
SMILES C1=CC(=CC=C1N)S(=O)(=O)O

Specifications

Melting Point ∽365°C
Density 1.485
Assay Percent Range 98%
Quantity 250 g
Beilstein 908765
Merck Index 14,8926
Solubility Information Slightly soluble in water. Insoluble in ethanol,ether,caustic soda and sodium carbonate.
Formula Weight 173.19
Percent Purity ≥98%
Chemical Name or Material Sulfanilic Acid
compliance-icons
Product Identifier
  • Sulfanilic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sulfanilic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.