missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide, MedChemExpress

Product Code. 16488428
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488428 10 g 10g
16400848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. Se returpolicy
Artikelnummer. 16488428 Leverandør Medchem Express Leverandørnr. HYB0242
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress Sulfanilamide is a competitive inhibitor for bacterial enzyme dihydropteroate synthetase with IC50 of 320 μM.

Sulfanilamide is a competitive inhibitor for bacterial enzyme dihydropteroate synthetase with IC50 of 320 μM.

Tekniske data

Chemical Name or Material Sulfanilamide
CAS 63-74-1
Color White
Purity Grade Notes Research
Molecular Formula C6H8N2O2S
Quantity 10 g
Synonym Sulphanilamide
Solubility Information DMSO : 100 mg/mL (580.72 mM; Need ultrasonic) ∣H2O : 10 mg/mL (58.07 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(N)C=C1)(N)=O
Molecular Weight (g/mol) 172.2
Formula Weight 172.2
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.