missing translation for 'onlineSavingsMsg'
Learn More

Sulfanilamide 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312972
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312972 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312972 Supplier Dr. Ehrenstorfer Supplier No. DREA17000000AL1000
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfanilamide
CAS 63-74-1
Analyte or Component Names Sulfanilamide
Shipping Condition +20°C
Molecular Formula C6 H8 N2 O2 S
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Sulfacetamide Sodium Imp. A (EP), Sulfacetamide Imp. A (EP), Sulfadiazine Imp. D (EP), Sulfadimidine Imp. D (EP), Sulfaguanidine Imp. A (EP), Sulfamethoxazole Imp. E (EP), 4-Aminobenzenesulfonamide, Sulfanilamide, 4-Aminobenzenesulphonamide, Sulphanilamide, Sulfadimidine Imp. D (Pharmeuropa), Sulfadiazine Imp. D (Pharmeuropa), (4-(Aminosulfonyl)phenyl)amine, 1162F, 4-(Aminosulfonyl)aniline, 4-Aminophenylsulfonamide, 4-Sulfamoylaniline, A-349, Albexan, Albosal, Ambeside, Antistrept, Astreptine, Astrocid, Bacteramid, Bactesid, Collomide, Colsulanyde, Copticide, Deseptyl, Dipron, Ergaseptine, Erysipan, Estreptocida, Exoseptoplix, F 1162, Fourneau 1162, Gerison, Gombardol, Infepan, Lusil, Lysococcine, NSC 7618, Neococcyl, Orgaseptine, PABS, Prontalbin, Prontosil Album, Prontosil I, Prontosil white, Prontylin, Pronzin Album, Proseptal, Proseptine, Proseptol, Pysococcine, Ro 1-3354, Rubiazol A, Sanamid, Septamide Album, Septanilam, Septinal, Septolix, Septoplex, Septoplix, Stopton Album, Stramid, Strepamide, Strepsan, Streptagol, Streptamid, Streptasol, Streptocid, Streptocid album, Streptocide, Streptocide (white), Streptoclase, Streptocom, Strepton, Streptopan, Streptosil, Streptozol, Streptozone, Sulfamidyl, Sulfamine, Sulfana, Sulfanalone, Sulfanidyl, Sulfanil, Sulfanilamide Vaginal Cream, Sulfocidine, Sulfonamide, Sulfonamide P, Sulfonylamide, Sulphonamide, Therapol, Tolder, White streptocide, p-Aminobenzenesulfonamide, p-Aminophenylsulfonamide, p-Anilinesulfonamide, p-Sulfamidoaniline, p-Sulfamoylaniline, Ph Eur Sulfamethoxazole Impurity E, Sulfanilamide, Sulfacetamide Sodium Impurity A, Sulfadiazine Impurity D, Sulfadimethoxine Impurity E, Sulfadimethoxine Sodium Impurity E, Sulfadimidine Impurity D, Sulfaguanidine Impurity A, Sulfamethoxazole Impurity E, Sulfacetamide Impurity A, Sulfadimethoxine Impurity E
InChI Formula InChI=1S/C6H8N2O2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H2,8,9,10)
SMILES Nc1ccc(cc1)S(=O)(=O)N
IUPAC Name 4-aminobenzenesulfonamide
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 172.2
Quantity 1 mL
Formula Weight 172.0306 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.