missing translation for 'onlineSavingsMsg'
Learn More

Sulfamoyl Chloride, TRC

Product Code. 30420882
Change view
Click to view available options
Quantity:
100 mg
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30420882 10 g
30410027 25 g
30413517 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30420882 Supplier Toronto Research Chemicals Supplier No. TRCS68902510G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-42-9
Molecular Formula Cl H2 N O2 S
Molecular Weight (g/mol) 115.54
Synonym Amidosulfonic Acid Chloride, Amidosulfonyl Chloride, Aminosulfonyl Chloride, Chloroimidosulfuric Acid, NSC 158265
IUPAC Name sulfamoyl chloride
SMILES NS(=O)(=O)Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/ClH2NO2S/c1-5(2,3)4/h(H2,2,3,4)
Formula Weight 114.9495
Chemical Name or Material Sulfamoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.