missing translation for 'onlineSavingsMsg'
Learn More

Sulfamonomethoxine, TRC

Product Code. 30138093
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138093 1 g 1g
17984805 100 mg 100mg
17903555 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138093 Supplier Toronto Research Chemicals Supplier No. TRCS6991451G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1220-83-3
Molecular Formula C11 H12 N4 O3 S
Molecular Weight (g/mol) 280.3
Synonym 4-Amino-N-(6-methoxy-4-pyrimidinyl)-benzenesulfonamide, Sulfanilamide, N1-(6-methoxy-4-pyrimidinyl)- (6CI,8CI), 4-Methoxy-6-sulfanilamidopyrimidine, 4-Sulfanilamido-6-methoxypyrimidine, 6-Methoxy-4-sulfanilamidopyrimidine, 6-Sulfanilamido-4-methoxypyrimidine, DJ 1550, DS 36, Daimeton, Duphadin, ICI 32525, N1-(6-Methoxy-4-pyrimidinyl)sulfanilamide, Ro 4-3476, Sulfamonomethoxin, Sulfamonomethoxine
IUPAC Name 4-amino-N-(6-methoxypyrimidin-4-yl)benzenesulfonamide
SMILES COc1cc(NS(=O)(=O)c2ccc(N)cc2)ncn1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H12N4O3S/c1-18-11-6-10(13-7-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7H,12H2,1H3,(H,13,14,15)
Formula Weight 280.063
Percent Purity >95%
Chemical Name or Material Sulfamonomethoxine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.