missing translation for 'onlineSavingsMsg'
Learn More

Sulfamonomethoxine, 98%

Product Code. 15478199
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15478199 250 mg 250mg
15468199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478199 Supplier Thermo Scientific Alfa Aesar Supplier No. J66733.MD
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Long-acting sulfonamide antibacterial agent. It is used to study degradation by Fe3O4 magnetic nanoparticles. It is used in blood kinetic studies and is used to study capsule formation of Bordetella bronchiseptica. | CAS: 1220-83-3 | C11H12N4O3S | 280.302 g/mol

Sulfamonomethoxine is a long acting sulfonamide antibacterial agent. It is used to study degradation by Fe3O4 magnetic nanoparticles. It is used in blood kinetic studies and is used to study capsule formation of Bordetella bronchiseptica.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1220-83-3
Molecular Formula C11H12N4O3S
Molecular Weight (g/mol) 280.302
MDL Number MFCD00063466
InChI Key WMPXPUYPYQKQCX-UHFFFAOYSA-N
Synonym sulfamonomethoxine, sulfamonomethoxin, daimeton, sulfamonometoxina, sulfamonomethoxinum, 4-amino-n-6-methoxypyrimidin-4-yl benzenesulfonamide, 4-amino-n-6-methoxy-4-pyrimidinyl benzenesulfonamide, benzenesulfonamide, 4-amino-n-6-methoxy-4-pyrimidinyl, n1-6-methoxy-4-pyrimidinyl sulfanilamide, sulfamonomethoxinum inn-latin
PubChem CID 5332
ChEBI CHEBI:32164
IUPAC Name 4-amino-N-(6-methoxypyrimidin-4-yl)benzenesulfonamide
SMILES COC1=NC=NC(=C1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Melting Point 203°C to 206°C
Boiling Point 513°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 250 mg
Beilstein 621128
Solubility Information Soluble in water at 10mg/ml with warming. Also soluble in ethanol or methanol
Formula Weight 280.3
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Sulfamonomethoxine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.