missing translation for 'onlineSavingsMsg'
Learn More

Sulfamide, TRC

Product Code. 30411552
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411552 1 g -
30422081 100 mg 100mg
30411577 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30411552 Supplier Toronto Research Chemicals Supplier No. TRCS6990791G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-58-9
Molecular Formula H4 N2 O2 S
Molecular Weight (g/mol) 96.11
Synonym Sulfamide, Imidosulfamic acid, NSC 252, Sulfamamide, Sulfonyl diamide, Sulfuric diamide, Sulfuryl amide, Sulfuryl diamide, Sulphamide
IUPAC Name sulfamide
SMILES NS(=O)(=O)N

Specifications

Quantity 1 g
InChI Formula InChI=1S/H4N2O2S/c1-5(2,3)4/h(H4,1,2,3,4)
Formula Weight 95.9993
Chemical Name or Material Sulfamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.