missing translation for 'onlineSavingsMsg'
Learn More

Sulfamide, 99%

Product Code. 11442968
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442968 50 g 50g
11422968 2.5 g 2.5g
11432968 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442968 Supplier Thermo Scientific Alfa Aesar Supplier No. A18680.18
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7803-58-9 | H4N2O2S | 96.10 g/mol

It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a carbonic anhydrase inhibitor. It is widely used as a pharmaceutical intermediate.

Solubility
Soluble in water, hot alcohol, acetone. Sparingly soluble in cold alcohol

Notes
Moisture Sensitive. Store away from oxidizing agents, water/humidity. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-58-9
Molecular Formula H4N2O2S
Molecular Weight (g/mol) 96.10
MDL Number MFCD00011606
InChI Key NVBFHJWHLNUMCV-UHFFFAOYSA-N
Synonym sulfuric diamide, sulfamamide, sulfuryl amide, sulfonyl diamide, sulfuryl diamide, sulfamoylamine, sulphamide, sulphuric diamide, unii-vs7tzw634v, imidosulfamic acid
PubChem CID 82267
ChEBI CHEBI:29368
SMILES NS(N)(=O)=O

Specifications

Melting Point 91°C to 92°C
Density 1.61
Quantity 50 g
Sensitivity Moisture sensitive
Merck Index 14,8922
Solubility Information Soluble in water, hot alcohol, acetone; Sparingly soluble in cold alcohol
IUPAC Name sulfamide
Formula Weight 96.11
Percent Purity 99%
Chemical Name or Material Sulfamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.