missing translation for 'onlineSavingsMsg'
Learn More

Sulfamide, 98+%

Product Code. 10328340 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10328340 25 g 25g
10183830 5 g 5g
11974391 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10328340 missing translation for 'mfr' Thermo Scientific missing translation for 'supplierNo' 199020250
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7803-58-9 | H4N2O2S | 96.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-58-9
Molecular Formula H4N2O2S
Molecular Weight (g/mol) 96.10
MDL Number MFCD00011606
InChI Key NVBFHJWHLNUMCV-UHFFFAOYSA-N
Synonym sulfuric diamide, sulfamamide, sulfuryl amide, sulfonyl diamide, sulfuryl diamide, sulfamoylamine, sulphamide, sulphuric diamide, unii-vs7tzw634v, imidosulfamic acid
PubChem CID 82267
ChEBI CHEBI:29368
IUPAC Name sulfamoylamine
SMILES NS(N)(=O)=O

Specifications

Melting Point 87.0°C to 91.0°C
Color White
Density 1.6100g/mL
pH 5.0 to 7.0 (10% soln. aq. soln.)
Elemental Analysis 98.0% min.
Loss on Drying 0.5% (50°C, 2 hrs) (vacuum) max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula H2NSO2NH2
Quantity 25 g
Merck Index 15,9054
Specific Gravity 1.61
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone,hot alcohol
Formula Weight 96.11
Percent Purity 98+%
Related Substances (TLC) Passes test
Physical Form Crystalline Powder
Chemical Name or Material Sulfamide
Show More Show Less
compliance-icons
Product Identifier
  • Sulfamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.