missing translation for 'onlineSavingsMsg'
Learn More

Sulfamidase/SGSH Antibody (1018331) [DyLight 488], Novus Biologicals™

Product Code. 30664796 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30664796 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30664796 Supplier Novus Biologicals Supplier No. FAB83801K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sulfamidase/SGSH Monoclonal antibody specifically detects Sulfamidase/SGSH in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sulfamidase/SGSH
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1018331
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias heparan sulfate sulfatase, HSSN-sulphoglucosamine sulphohydrolase, MPS3Asulphamidase, N-sulfoglucosamine sulfohydrolase, SFMDEC 3.10.1.1, Sulfoglucosamine sulfamidase, Sulphamidase
Host Species Mouse
Immunogen Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Sulfamidase/SGSH, Arg23-Leu502, Accession # P51688
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6448
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.