missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, ACS Reagent Grade, Ricca Chemical

Product Code. 31749308
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Natural Poly Bottle
White Poly Wide Mouth Bottle
Unit Size:
100g
2.50 L
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
31749308 500 g White Poly Wide Mouth Bottle 500g
31749934 100 g White Poly Wide Mouth Bottle 100g
31750644 2.5 kg Natural Poly Bottle 2.50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31749308 Supplier Ricca Chemical Company Supplier No. RDCS1100500B1
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid

Specifications

CAS Min % 98.0
Color White
CAS Max % 100.0
Heavy Metals (as Pb) max. 0.001%
Packaging White Poly Wide Mouth Bottle
Residue after Ignition max. 0.01%
Quantity 500 g
Physical Form Crystals
Synonym amidosulfonic acid, amidosulfuric acid, aminosulfonic acid, imidosulfonic acid, sulfamidic acid, sulfaminic acid, sulphamic acid, jumbo, aminosulfuric acid, sulphamidic acid
SMILES NS(=O)(=O)O
Molecular Weight (g/mol) 97.088
Percent Purity 99.3 to 100.3%
Grade ACS
Chemical Name or Material Sulfamic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.