missing translation for 'onlineSavingsMsg'
Learn More

Sulfamic Acid, ACS Reagent Grade, Ricca Chemical

Product Code. 31750644
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Natural Poly Bottle
White Poly Wide Mouth Bottle
Unit Size:
100g
2.50 L
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
31750644 2.5 kg Natural Poly Bottle 2.50 L
31749934 100 g White Poly Wide Mouth Bottle 100g
31749308 500 g White Poly Wide Mouth Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31750644 Supplier Ricca Chemical Company Supplier No. RDCS11002.5A5
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 5329-14-6
Molecular Formula H3NO3S
InChI Key IIACRCGMVDHOTQ-UHFFFAOYSA-N
PubChem CID 5987
ChEBI CHEBI:9330
IUPAC Name sulfamic acid

Specifications

Color White
CAS Min % 98.0
CAS Max % 100.0
Heavy Metals (as Pb) max. 0.001%
Packaging Natural Poly Bottle
Residue after Ignition max. 0.01%
Quantity 2.5 kg
Physical Form Crystals
Synonym amidosulfonic acid
SMILES NS(O)(=O)=O
Molecular Weight (g/mol) 97.09
Percent Purity 99.3 to 100.3%
Grade ACS
Chemical Name or Material Sulfamic Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.