missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxypyrazine, TRC

Product Code. 30010312
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010312 10 mg 10mg
30006372 25 mg 25mg
30007425 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010312 Supplier Toronto Research Chemicals Supplier No. TRCS69923010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152-47-6
Molecular Formula C11 H12 N4 O3 S
Molecular Weight (g/mol) 280.3
Synonym Benzenesulfonamide, 4-amino-N-(3-methoxypyrazinyl)- (9CI), Sulfanilamide, N1-(3-methoxypyrazinyl)- (8CI), 2-Methoxy-3-sulfanilamidopyrazine, 2-Sulfanilamido-3-methoxypyrazine, 3-Methoxy-2-sulfanilamidopyrazine, 3-Methoxy-2-sulfapyrazine, 3-Methoxypyrazine sulfanilamide, AS 18908, Dalysep, F.I. 5978, Farmitalia 204/122, Kelfizin, Kelfizina, Kelfizine W, Longum, N1-(3-Methoxy-2-pyrazinyl)sulfanilamide, NSC 110433, Policydal, Polycidal, SMP2, Sulfalene, Sulfamethopyrazine, Sulfamethoxypyrazine, Sulfametopyrazine, Sulfapyrazinemethoxine, Sulfapyrazinemethoxyine, Sulfapyrazinemethoxyne, Vetkelfizina
IUPAC Name 4-amino-N-(3-methoxypyrazin-2-yl)benzenesulfonamide
SMILES COc1nccnc1NS(=O)(=O)c2ccc(N)cc2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C11H12N4O3S/c1-18-11-10(13-6-7-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7H,12H2,1H3,(H,13,15)
Formula Weight 280.063
Chemical Name or Material Sulfalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.