missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfamethoxazole N4-beta-D-Glucoside, Mikromol™
Description
Sulfamethoxazole N4-beta-D-Glucoside
Specifications
Specifications
| Chemical Name or Material | Sulfamethoxazole N4-glucoside |
| CAS | 119691-75-7 |
| Analyte or Component Names | Sulfamethoxazole N4-glucoside |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H21 N3 O8 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Sulfamethoxazole N4-glucoside, 4-(β-D-glucopyranosylamino)-N-(5-methyl-3-isoxazolyl)-benzenesulfonamide |
| InChI Formula | InChI=1S/C16H21N3O8S/c1-8-6-12(18-27-8)19-28(24,25)10-4-2-9(3-5-10)17-16-15(23)14(22)13(21)11(7-20)26-16/h2-6,11,13-17,20-23H,7H2,1H3,(H,18,19)/t11-,13-,14+,15-,16-/m1/s1 |
| SMILES | Cc1onc(NS(=O)(=O)c2ccc(N[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)cc2)c1 |
| IUPAC Name | N-(5-methyl-1,2-oxazol-3-yl)-4-[[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]amino]benzenesulfonamide |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?