missing translation for 'onlineSavingsMsg'
Learn More

Sulfamethoxazole, 100%, MP Biomedicals™

Product Code. 11445131 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11445131 100 g 100g
11435131 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445131 Supplier MP Biomedicals Supplier No. 0215671180

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 723-46-6
Molecular Formula C10H11N3O3S
Molecular Weight (g/mol) 253.276
InChI Key JLKIGFTWXXRPMT-UHFFFAOYSA-N
Synonym sulfamethoxazole, sulphamethoxazole, sulfisomezole, gantanol, sulfamethoxazol, metoxal, sulfamethylisoxazole, simsinomin, radonil, sinomin
PubChem CID 5329
ChEBI CHEBI:9332
IUPAC Name 4-amino-N-(5-methyl-1,2-oxazol-3-yl)benzenesulfonamide
SMILES CC1=CC(=NO1)NS(=O)(=O)C2=CC=C(C=C2)N

Specifications

Color White
Quantity 100 g
Formula Weight 253.3
Physical Form Powder
Chemical Name or Material Sulfamethoxazole
compliance-icons
Product Identifier
  • sulphamethoxazole
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: sulphamethoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.