missing translation for 'onlineSavingsMsg'
Learn More

Sulfamerazine, TRC

Product Code. 30138087
Change view
Click to view available options
Quantity:
50 mg
1 g
50 g
Unit Size:
1g
50g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138087 50 g 50g
30135072 1 g 1g
17972965 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138087 Supplier Toronto Research Chemicals Supplier No. TRCS68900050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-79-7
Molecular Formula C11 H12 N4 O2 S
Molecular Weight (g/mol) 264.3
Synonym 4-Amino-N-(4-methyl-2-pyrimidyl)benzenesulfonamide, Sulfamerazine, Sulfadimidine Imp. A (Pharmeuropa)
IUPAC Name 4-amino-N-(4-methylpyrimidin-2-yl)benzenesulfonamide
SMILES Cc1ccnc(NS(=O)(=O)c2ccc(N)cc2)n1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C11H12N4O2S/c1-8-6-7-13-11(14-8)15-18(16,17)10-4-2-9(12)3-5-10/h2-7H,12H2,1H3,(H,13,14,15)
Formula Weight 264.0681
Percent Purity >95%
Chemical Name or Material Sulfamerazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.