missing translation for 'onlineSavingsMsg'
Learn More

Sulfaguanidine 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30327109
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30327109 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30327109

Brand: Dr. Ehrenstorfer DREA16990675AL1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfaguanidine
CAS 57-67-0
Analyte or Component Names Sulfaguanidine
Shipping Condition +20°C
Molecular Formula C7 H10 N4 O2 S
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Sulfadimidine Imp. C (EP), Sulfamethazine USP Related Compound A, Sulfamethazine USP RC A, (4-Aminophenylsulfonyl)guanidine, Sulfaguanidine, Sulfadimidine Imp. C (Pharmeuropa), N1-(Diaminomethylene)sulfanilamide, 1-Sulfanilylguanidine, 1-[(p-Aminophenyl)sulfonyl]guanidine, 4-Amino-N-(aminoiminomethyl)benzenesulfonamide, 4-Aminobenzenesulfonylguanidine, 4-Aminophenylsulfonylguanidine, A 307, Abiguanil, Aterian, Diacta, Emerin (pharmaceutical), Ganidan, Guamide, Guanicil, Guanidan, N-Guanylsulfanilamide, N1-Amidinosulfanilamide, N1-Guanidylsulfanilamide, N1-Guanylsulfanilamide, NSC 14041, Orgaguanidon, RP 2275, Resulfon, Ruocid, Shigatox, Suganyl, Sulfaguanidin, Sulfaguanil, Sulfaguine, Sulfanilguanidine, Sulfanilylguanidine, Sulfentidine, Sulfoguanidine, Sulfoguanil, Sulfoguanyl, Sulfoguenil, Sulgin, Sulphaguanidine, [(p-Aminophenyl)sulfonyl]guanidine, p-Aminobenzenesulfoguanidide, p-Aminobenzenesulfonylguanidine, Sulfamethazine Related Compound A, Sulfadiazine Impurity C, Sulfadimidine Impurity C
InChI Formula InChI=1S/C7H10N4O2S/c8-5-1-3-6(4-2-5)14(12,13)11-7(9)10/h1-4H,8H2,(H4,9,10,11)
SMILES NC(=N)NS(=O)(=O)c1ccc(N)cc1
IUPAC Name 1-(4-aminophenyl)sulfonylguanidine
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 214.24
Quantity 1 mL
Formula Weight 214.0524 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.