missing translation for 'onlineSavingsMsg'
Learn More

Sulfadoxine, TRC

Product Code. 30138565
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138565 10 g 10g
30136126 1 g 1g
17942525 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138565 Supplier Toronto Research Chemicals Supplier No. TRCS69907010G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2447-57-6
Molecular Formula C12 H14 N4 O4 S
Molecular Weight (g/mol) 310.33
Synonym Sulfadoxine, 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide, Benzenesulfonamide, 4-amino-N-(5,6-dimethoxy-4-pyrimidinyl)-, Sulfanilamide, N1-(5,6-dimethoxy-4-pyrimidinyl)-, 4,5-Dimethoxy-6-sulfanilamidopyrimidine, 4-Amino-N-(5,6-dimethoxy-4-pyrimidinyl)benzensulfonamide, 4-Sulfanilamido-5,6-dimethoxypyrimidine, Fanasil, Fanasulf, N1-5,6-Dimethoxy-4-pyrimidinylsulfanilamide, Orthosulfin, Ro 4-4393, Sanasil, Sulfadoxin, Sulfadoxine, Sulformethoxine, Sulformetoxin, Sulformetoxine, Sulforthodimethoxine, Sulforthomidine, WR 4073
IUPAC Name 4-amino-N-(5,6-dimethoxypyrimidin-4-yl)benzenesulfonamide
SMILES COc1ncnc(NS(=O)(=O)c2ccc(N)cc2)c1OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C12H14N4O4S/c1-19-10-11(14-7-15-12(10)20-2)16-21(17,18)9-5-3-8(13)4-6-9/h3-7H,13H2,1-2H3,(H,14,15,16)
Formula Weight 310.0736
Percent Purity >95%
Chemical Name or Material Sulfadoxine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.